Learn More
Sklar Instruments Surgi-OR Adson Dressing Forceps, Sklar 4-3/4 in., Blunt, Straight, Serrated, Delicate, Stainless Steel. EA.
SDP
Catalog No. 501184235 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Adson Dressing Forceps, Sklar. 4-3/4 in., Blunt, Straight, Serrated, Delicate, Stainless Steel. Surgi-OR Adson Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Surgi-OR mid-grade instruments provide cost-effective, reliable instrumentation for the operating room and surgical centers. This product is straight with delicate, serrated tips and a length of 4-3/4 inches.
Catalog No. 50-118-4235 Supplier Sklar Instruments Supplier No. 95772
Surgi-OR Adson Dressing Forceps, Sklar. 4-3/4 in., Blunt, Straight, Serrated, Delicate, Stainless Steel. Surgi-OR Adson Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Surgi-OR mid-grade instruments provide cost-effective, reliable instrumentation for the operating room and surgical centers. This product is straight with delicate, serrated tips and a length of 4-3/4 inches.