Learn More
Sklar Instruments Surgi-OR Tissue Forceps, Sklar 5 in., Straight, 1x2 Teeth, Stainless Steel. EA.
SDP
Catalog No. 501184234 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Tissue Forceps, Sklar. 5 in., Straight, 1x2 Teeth, Stainless Steel. Surgi-OR Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. This product is straight with 1x2 teeth.
Catalog No. 50-118-4234 Supplier Sklar Instruments Supplier No. 95757
Surgi-OR Tissue Forceps, Sklar. 5 in., Straight, 1x2 Teeth, Stainless Steel. Surgi-OR Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures. These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. This product is straight with 1x2 teeth.