Learn More
Sklar Instruments Surgi-OR TC Webster Needle Holder, Sklar 5-1/4 in., Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA.
SDP
Catalog No. 501184165 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR TC Webster Needle Holder, Sklar. 5-1/4 in., Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Surgi-OR Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth tips and a length of 5-1/4 inches.
Catalog No. 50-118-4165 Supplier Sklar Instruments Supplier No. 95130
Surgi-OR TC Webster Needle Holder, Sklar. 5-1/4 in., Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Surgi-OR Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth tips and a length of 5-1/4 inches.