Learn More
Med Vet International Surgibond Vet Use Tissue Adhesive, 2.5gr Bottle, Each
Catalog No. 502829765
Encompass_Preferred
Surgibond contains n-butyl cyanoacrylate tissue adhesive and is used to bond tissue together for veterinary procedures (i. e. cat declaws, lacerations, abrasions, suture/staple line sealing, tooth extractions; oral surgeries). Surgibond may be applied for the closure of wounds after careful adaption of the wound edges as a suture substitute, for additional suture support and for protection of infections after sutures. Surgibond contains blue dye to allow user to easily see where product has been applied. On contact with body tissue, Surgibond adhesive changes from liquid to solid state by polymerizing within seconds to seal the wound. Adhesive stops minor bleeding and binds wound edges. May eliminate need for sutures/b and ages in some instances Acts as a barrier to keep foreign matter from entering the wound Tissue adhesive is sloughed as healing occurs Comes with 5 applicators
Catalog No. 50-282-9765 Supplier Med Vet International Supplier No. ZG2
Surgibond contains n-butyl cyanoacrylate tissue adhesive and is used to bond tissue together for veterinary procedures (i. e. cat declaws, lacerations, abrasions, suture/staple line sealing, tooth extractions; oral surgeries). Surgibond may be applied for the closure of wounds after careful adaption of the wound edges as a suture substitute, for additional suture support and for protection of infections after sutures. Surgibond contains blue dye to allow user to easily see where product has been applied. On contact with body tissue, Surgibond adhesive changes from liquid to solid state by polymerizing within seconds to seal the wound. Adhesive stops minor bleeding and binds wound edges. May eliminate need for sutures/b and ages in some instances Acts as a barrier to keep foreign matter from entering the wound Tissue adhesive is sloughed as healing occurs Comes with 5 applicators