Learn More
Braintree Scientific Surgical Board, 8.125 x 16""
SDP
Catalog No. 501954076
Encompass
Surgical Board, 8.125 x 16" Animal Range: up to 12" long Surgery boards are formed of one piece high-temp NorylTM for durability and ease of cleaning. Five adjustable Nylon cords are included. All rodent restrainersare manufactured from clear materials to provide complete visual awareness of the state of your animal. Additional sizes andstyles available. Call for details. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4076 Supplier Braintree Scientific Supplier No. 510SS
Surgical Board, 8.125 x 16" Animal Range: up to 12" long Surgery boards are formed of one piece high-temp NorylTM for durability and ease of cleaning. Five adjustable Nylon cords are included. All rodent restrainersare manufactured from clear materials to provide complete visual awareness of the state of your animal. Additional sizes andstyles available. Call for details. **Sold for research purposes only and not for human use**