Learn More
Surgical Design No. 10 Carbon Scalpel Blade
Catalog No. 22079690 Shop All Surgical Design Products
This item is not returnable. View return policy
Catalog No. 22-079-690 Supplier Surgical Design Supplier No. BLADE#10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Traditional curved cutting edge blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • Used for making small incisions in skin and muscle
  • Used in more specialized surgeries such as for harvesting radial artery during a coronary artery bypass operation, opening bronchus during thoracic surgery and for Inguinal hernia repair
Specifications
For Use With (Equipment) Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L