Learn More
Surgical Design No. 11 Carbon Scalpel Blade
Catalog No. 22079691 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 11
Catalog No. Size
22-079-691 # 11
1 options
This item is not returnable. View return policy
Catalog No. 22-079-691 Supplier Surgical Design Supplier No. BLADE#11
Add to Cart
Add to Cart
This item is not returnable. View return policy
Elongated triangular blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • Sharpened along hypotenuse edge
  • Strong pointed tip making it ideal for stab incisions
Specifications
Size # 11
For Use With (Equipment) Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L