Learn More
Surgical Design No. 12 Small Pointed Stainless Steel Scalpel Blade
Catalog No. 22079700 Shop All Surgical Design Products
This item is not returnable. View return policy
Catalog No. 22-079-700 Supplier Surgical Design Supplier No. BLADE#12S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Crescent shaped blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • Sharpened along inside edge of curve
  • Used for surgical incision of an artery
  • Used for calculus removal by incision of ureter
  • Used for surgical incision of renal pelvis of kidney for removal of kidney stone
Specifications
For Use With (Equipment) Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L