Learn More
Surgical Design No. 15 Small Stainless Steel Scalpel Blade
Catalog No. 22079701 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 15
Catalog No. Size
22-079-701 # 15
1 options
Catalog No. 22-079-701 Supplier Surgical Design Supplier No. BLADE#15S
Add to Cart
Add to Cart
Small curved cutting edge blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • Ideal for making short and precise incisions
  • Used for excision of skin lesion or recurrent sebaceous cyst and for opening coronary arteries
Specifications
Size # 15
For Use With (Equipment) Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L