Learn More
Surgical Design No. 15C Long Stainless Steel Scalpel Blade
Catalog No. 22079702 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 15
Catalog No. Size
22-079-702 # 15
1 options
This item is not returnable. View return policy
Catalog No. 22-079-702 Supplier Surgical Design Supplier No. BLADE#15CS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Long, extended cutting edge blade fits handles 3, 3L, 3 Graduated, 5B, 7, 9, B3 and B3L
  • More extended cutting edge than traditional No.15 blade
  • Provides additional reach for dentist carrying out periodontal procedures
  • Used by Podiatrists for nucleating corns around webbing of toes
Specifications
Size # 15
For Use With (Equipment) Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L