Learn More
Surgical Design No. 20 Large Stainless Steel Scalpel Blade
Catalog No. 22079703 Shop All Surgical Design Products
Catalog No. 22-079-703 Supplier Surgical Design Supplier No. BLADE#20S
Add to Cart
Add to Cart
Curved cutting edge blade fits handles 4, 4L, 4 Graduated and 6B
  • Large version of no.10 blade
  • Flat, unsharpened back edge
  • Used for orthopaedic and general surgical procedures
Specifications
For Use With (Equipment) Handles # 4, # 4L, # 4 Graduated and # 6B