Learn More
Surgical Design No. 21 Carbon Scalpel Blade
Catalog No. 22079696 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 21
Catalog No. Size
22-079-696 # 21
1 options
This item is not returnable. View return policy
Catalog No. 22-079-696 Supplier Surgical Design Supplier No. BLADE#21
Add to Cart
Add to Cart
This item is not returnable. View return policy
Curved cutting blade fits handles 4, 4L, 4 Graduated and 6B
  • Larger version of No.10 blade
  • Flat, unsharpened back edge
  • Larger than No.20 but smaller than No.22
Specifications
Size # 21
For Use With (Equipment) Handles # 4, # 4L, # 4 Graduated and # 6B