Learn More
Surgical Design No. 22 Carbon Scalpel Blade
Catalog No. 22079697 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 22
Catalog No. Size
22-079-697 # 22
1 options
This item is not returnable. View return policy
Catalog No. 22-079-697 Supplier Surgical Design Supplier No. BLADE#22
Add to Cart
Add to Cart
This item is not returnable. View return policy
Curved cutting edge blade fits handles 4, 4L, 4 Graduated and 6B
  • Larger version of No.10 blade
  • Flat, unsharpened back edge
  • Used for skin incisions in both cardiac and thoracic surgrey and to cut bronchus in lung resection surgery
  • Sterile; individually wrapped
Specifications
Size # 22
For Use With (Equipment) Handles # 4, # 4L, # 4 Graduated and # 6B
Sterility Sterile