Learn More
Surgical Design No.23 Leaf-Shaped Stainless Steel Scalpel Blade
Catalog No. 22079705 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 23
Catalog No. Size
22-079-705 # 23
1 options
This item is not returnable. View return policy
Catalog No. 22-079-705 Supplier Surgical Design Supplier No. BLADE#23S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Leaf-shaped blade fits handles 4, 4L, 4 Graduated and 6B
  • Ssharpened along leading edge
  • For making long incisions such as an upper midline incision of abdomen during repair of perforated gastric ulcer
Specifications
Size # 23
For Use With (Equipment) Handles # 4, # 4L, # 4 Graduated and # 6B