Learn More
Surgical Design No. 70 Carbon Scalpel Blades for Post-Mortem, Autopsy and Research Use
Catalog No. 22079699 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 70
Catalog No. Size
22-079-699 # 70
1 options
This item is not returnable. View return policy
Catalog No. 22-079-699 Supplier Surgical Design Supplier No. BLADE#70
Add to Cart
Add to Cart
This item is not returnable. View return policy
Surgical Design™ Royaltek™ Post Mortem Autopsy Scalpel Blades are made from surgical carbon steel blades and sharpened to perfection for a super clean cut.
Surgical Design™ presents Royaltek™- our premium line of surgical blades
  • Rounded tip reduces puncture risk
  • Thicker blades reduce breakage
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical-quality blades
  • Individually sealed
  • Peelable packing
  • Sterile
  • Fits no.8 scalpel handle 
Note:  For post-mortem, autopsy and research use.
Specifications
For Use With (Application) For Postmortem, Autopsy and Research use
Size # 70
For Use With (Equipment) Handles # 4, # 4L, # 4 Graduated and # 6B