Learn More
Sklar Instruments Econo Surgical Knife Handle, Sklar, Straight, Stainless Steel. Econo #4 Scalpel Handle is similar to the #3 scalpel handle, but compatible with the larger size 20 series disposable blades. It is used for sharp dissection. PK/12.
SDP
Catalog No. 501437519 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Surgical Knife Handle, Sklar, Straight, Stainless Steel. Econo #4 Scalpel Handle is similar to the #3 scalpel handle, but compatible with the larger size 20 series disposable blades. It is used for sharp dissection. There are centimeter and millimeter markings on the handle for measuring tissue and a center groove for thumb and forefinger gripping. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.
This item is not returnable. View return policy
Catalog No. 50-143-7519 Supplier Sklar Instruments Supplier No. 21038
Econo Surgical Knife Handle, Sklar, Straight, Stainless Steel. Econo #4 Scalpel Handle is similar to the #3 scalpel handle, but compatible with the larger size 20 series disposable blades. It is used for sharp dissection. There are centimeter and millimeter markings on the handle for measuring tissue and a center groove for thumb and forefinger gripping. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.