Learn More
Surgical Design Surgical Knife Scalpel Handle
Catalog No. 22079767 Shop All Surgical Design Products
Change view
Click to view available options
Blade Material:
Plastic
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Material
22-079-767 Stainless Steel
22-079-768 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-079-767 Supplier Surgical Design Supplier No. 27108
Add to Cart
Add to Cart
Surgical Design™ Surgical Knife Scalpel Handle used for sharp and precise dissection. Compatible with 60 and 70 Series blades.
  • Surgical Design™ Premium products exceed performance and quality expectations. Guaranteed.
  • Handles ergonomically designed for precision work
  • Comfortable, reusable
  • Blades slide on smoothly and securely
  • Superb quality and workmanship
  • Premium surgical grade stainless steel
  • Corrosion protected by passivation and ultrasonic cleaning
  • Sterilizable by EtOH/Gamma Radiation
  • Fully autoclavable
Specifications
Blade Material Stainless Steel
Product Type Ergonomic Scalpel Handle
Size # 8
For Use With (Equipment) Compatible with # 60 and # 70 Series Blade
Disposable Reusable
Autoclavable Autoclavable