Learn More
Stoelting™ Surgical Lubricant
SDP
Catalog No. 10000693 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
2 oz.
4 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-000-693 2 oz.
10-000-694 4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-693 Supplier Stoelting™ Supplier No. 50976
Add to Cart
Add to Cart
Stoelting™ Surgical Lubricant forms a transparent lubricating film that spreads readily.
  • Non-irritating to tissue
  • Allows easy insertion of probes, catheters, surgical instruments, etc.
  • Sterile and bacteriostatic
Specifications
For Use With (Application) Neuroscience
Size 2 oz.
Includes Box of 12