Learn More
Med Vet International Surgical Neutral Detergent, J0647, 1 Gallon
Catalog No. 5000581007
Encompass_Preferred
Ultrasonic andmanual cleaning detergent for surgical instruments.• Very concentrated. 5ml makes 1 gallon• Works on metals, rubber, plastic and glass• High suds with rust inhibitor• Dispenser pump included• Available 4 gallons/case
Catalog No. 50-005-81007 Supplier Med Vet International Supplier No. JOR647
Ultrasonic andmanual cleaning detergent for surgical instruments.• Very concentrated. 5ml makes 1 gallon• Works on metals, rubber, plastic and glass• High suds with rust inhibitor• Dispenser pump included• Available 4 gallons/case