Learn More
Fine Science Tools Surgical Scissors, Sharp, Stainless Steel, 12 cm
SDP
Catalog No. NC1079624
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 41mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No
Catalog No. NC1079624 Supplier Fine Science Tools Supplier No. 1400512
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 41mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No