Learn More
Fine Science Tools Surgical Scissors, Sharp, Stainless Steel, 14.5 cm
SDP
Catalog No. NC9617856
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No
Catalog No. NC9617856 Supplier Fine Science Tools Supplier No. 1400214
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No