Learn More
Fine Science Tools Surgical Scissors, Tungsten Carbide, 14.5 cm
SDP
Catalog No. NC9556883
Encompass
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Tungsten Carbide
Catalog No. NC9556883 Supplier Fine Science Tools Supplier No. 1450114
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 47mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Tungsten Carbide