Learn More
Fine Science Tools Surgical Scissors, Blunt, Stainless Steel, 20 cm
SDP
Catalog No. NC0431919
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 60mm/ Alloy / Material: Stainless Steel/ Length: 20cm/ Serrated: No
Catalog No. NC0431919 Supplier Fine Science Tools Supplier No. 1400020
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 60mm/ Alloy / Material: Stainless Steel/ Length: 20cm/ Serrated: No