Learn More
Fine Science Tools Skin Marker, 0.02 mm Versatile Tip
SDP
Catalog No. NC9192981
Encompass
Product: Skin Marker with FST Ruler/ Features: Versatile tip for making fine (0.02mm) lines or a broad (2mm) line when held at an angle. The ink is violet, non toxic, non tattooing and surgical prep resistant/ Unit : Each
Catalog No. NC9192981 Supplier Fine Science Tools Supplier No. 1800030
Product: Skin Marker with FST Ruler/ Features: Versatile tip for making fine (0.02mm) lines or a broad (2mm) line when held at an angle. The ink is violet, non toxic, non tattooing and surgical prep resistant/ Unit : Each