Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Surgical Tweezers 3LUSO.S, 120 mm, Inox
SDP

Supplier:  Electron Microscopy Sciences 7892003

Encompass_Preferred

Medical tweezers are handcrafted to perfect tip symmetry and balance and the surface has an electropolish finish. With high precision fine tips and Dot Serrated handles for a perfect grip. All of these tweezers can be sterilized and they are all made from Inox.
OAL: 120 mm
Material: Inox
Tip width: B:0.10 mm
Tip thickness: C: 0.10 mm

Catalog No. 50-192-7221


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.