Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MediPurpose™ SurgiLance™ Lite Safety Lancet
SDP

Catalog No. 22000264
Change view
Click to view available options
Color:
Lime green
Purple
For Use With (Application):
Glucose test
Glucose test; Hematocrit tube
Depth (Metric):
1.8mm
2.2mm
Flow Type:
Low (10 to 20μL)
Medium (20 to 40μL)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Application) Depth (Metric) Flow Type
22-000-264 Lime green Glucose test; Hematocrit tube 2.2mm Medium (20 to 40μL)
22-000-263 Purple Glucose test 1.8mm Low (10 to 20μL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-264 Supplier MediPurpose™ Supplier No. SLL21G220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MediPurpose™ SurgiLance™ Lite safety lancets for capillary blood collection fingerstick users have an eco-friendly design that is ideal for small hands

  • Easy to use
  • Minimizes pain
  • Safe, single-use design
  • 21 & 28 guage needles
  • Eco-friendly design
  • Easier for small hands

 

Specifications

Disposable Single-use
Product Type Fingerstick Lancet
Flow Type Medium (20 to 40μL)
For Use With (Application) Glucose test; Hematocrit tube
Needle Gauge G21
Style Lite
Color Lime green
Depth (Metric) 2.2mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.