Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MediPurpose™ SurgiLance™ Lite Safety Lancets
SDP

Catalog No. 22000234
Change view
Click to view available options
Depth (English) Needle:
1.8mm
Color:
Gray
Flow Type:
Low (5 to 10μL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English) Needle Color Flow Type
22-000-234 1.8mm Gray Low (5 to 10μL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-000-234 Supplier MediPurpose™ Supplier No. SLL180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for users with small hands.

  • Eco-friendly design
  • Single use design
  • 28-gauge needle
  • Easy to use
  • Minimizes pain

Specifications

Disposable Single-use
Model SurgiLance™ Lite
Depth (English) Needle 1.8mm
Flow Type Low (5 to 10μL)
For Use With (Application) Glucose test
Color Gray
Needle Gauge 28G
Product Type Safety Lancet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.