Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MediPurpose™ SurgiLance™ Safety Lancet
SDP

Catalog No. 22000262
Change view
Click to view available options
Needle Gauge:
G21
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-000-262 G21
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-000-262 Supplier MediPurpose™ Supplier No. SLN100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MediPurpose™ SurgiLance™ safety lancets for capillary blood collection fingerstick users

  • Easy to use
  • Minimizes pain
  • Safe, single-use design
  • 21 guage needles
  • 18 guage blades

Specifications

Disposable Single-use
Product Type Fingerstick Lancet
Flow Type Low (5 to 10μL)
Color Yellow
Depth (Metric) 1.0mm
For Use With (Application) Glucose test; Geriatrics
Style Classic
Needle Gauge G21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.