Learn More
Leica Biosystems SURGIPATH ACID ALCOHOL G
Catalog No. NC1493549
Encompass_Preferred
Acid Alcohol is used in histology and cytology to remove excess stain and define nuclei following hematoxylin staining in the H&E and PAP methods. In order to produce a good H&E stain with crisp nuclear detail, it is critical that certain Hematoxylins are differentiated correctly, making Acid Alcohol a highly effective solution. Available with 1% or 0.5% hydrochloric acid in 70% reagent grade alcohol, this Acid Alcohol can greatly improve reproducibility of staining results and help achieve standardization.
Catalog No. NC1493549 Supplier Leica Biosystems Supplier No. 3803650
Acid Alcohol is used in histology and cytology to remove excess stain and define nuclei following hematoxylin staining in the H&E and PAP methods. In order to produce a good H&E stain with crisp nuclear detail, it is critical that certain Hematoxylins are differentiated correctly, making Acid Alcohol a highly effective solution. Available with 1% or 0.5% hydrochloric acid in 70% reagent grade alcohol, this Acid Alcohol can greatly improve reproducibility of staining results and help achieve standardization.