Learn More
Kent Scientific Corporation SURGISUITE MULTI-FUNCTNL,MICE
SDP
Catalog No. NC2157315 Shop All Kent Scientific Products
Encompass
SurgiSuite Multi-Functional Surgical Platform for Mice, with Lighting and Magnification, Warming, and Intubation. The SurgiSuite Multi-Functional Surgical Platform has an easy-to-clean, chemical resistant base which includes a channel for collecting fluids during surgical operations. Integrated Far Infrared Warming is designed into the platform.
This item is not returnable. View return policy
Catalog No. NC2157315 Supplier Kent Scientific Corporation Supplier No. SURGIM04
SurgiSuite Multi-Functional Surgical Platform for Mice, with Lighting and Magnification, Warming, and Intubation. The SurgiSuite Multi-Functional Surgical Platform has an easy-to-clean, chemical resistant base which includes a channel for collecting fluids during surgical operations. Integrated Far Infrared Warming is designed into the platform.