Learn More
Med Vet International Surgivet, Guttural Pouch, Endoscopic, Flushing, Catheter, 6Fr X 250Cm
Catalog No. 502828195
Encompass_Preferred
PTFE catheter used for lung lavage or introduction into the guttural pouch to flush out debris. Introduction is aided by visual indicators at 2.5, 5, and 7.5 cm from the distal tip. Sterile. Avian | canine | cattle | equine | feline | goat | poultry | sheep | swine
Catalog No. 50-282-8195 Supplier Med Vet International Supplier No. PHVENDO250
PTFE catheter used for lung lavage or introduction into the guttural pouch to flush out debris. Introduction is aided by visual indicators at 2.5, 5, and 7.5 cm from the distal tip. Sterile. Avian | canine | cattle | equine | feline | goat | poultry | sheep | swine