Learn More
TARGETMOL CHEMICALS INC SURICLONE 5MG
SDP
Catalog No. 502580483
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Suriclone (RP 31264) is a selective cyclic pyrrolidone analog with sedative and anxiolytic activity but not significant sedative effects.Suriclone shows its effects by modulating GABA-A receptors. purity: 98%
Catalog No. 50-258-0483 Supplier TARGETMOL CHEMICALS INC Supplier No. T288875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Suriclone (RP 31264) is a selective cyclic pyrrolidone analog with sedative and anxiolytic activity but not significant sedative effects.Suriclone shows its effects by modulating GABA-A receptors. purity: 98%