Learn More
Surrogate Fortification Standard, EPA 525.2, Multi Component Mixture, 1 mL
Specifications
Specifications
| Chemical Name or Material | Surrogate Standard Fortification Solution |
| Name Note | 500μg/mL |
| CAS Max % | 99.75, 0.06, 0.06, 0.06, 0.06 |
| CAS | 81-20-9, 1520-96-3, 115-86-6, 1718-52-1, 67-64-1 |
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 1 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 500μg/mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.