Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surrogate Standard Mixture, 4000μg/mL in methylene chloride, Ultra Scientific

Catalog No. USISM3311
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ISM-3311 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ISM-3311 Supplier Agilent Technologies Supplier No. ISM3311
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Surrogate Standard Mixture
CAS 75-09-2, 1718-51-0, 367-12-4, 4165-60-0, 4165-62-2
CAS Max % 98.2, 0.3, 0.3, 0.3, 0.3, 0.3
Analyte or Component Names Methylene chloride,2-fluorobiphenyl,2-fluorophenol
Element Methylene Chloride, 2-Fluorobiphenyl, 2-Fluorophenol
Solvent or Matrix Methylene Chloride
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 4000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.