Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surrogate Standard Mixture (For EPA 601, 624, 8010A and B)

Catalog No. USSTM290N1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-STM-290N-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-STM-290N-1 Supplier Agilent Technologies Supplier No. STM290N1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Page 303C, Designates A2LA-Certified product

Specifications

Chemical Name or Material Surrogate Standard Mixture
Packaging Ampule
Molecular Formula CH4O
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methanol:
Bromochloromethane
2-Bromo-1-chloropropane
1, 4-Dichlorobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.