Learn More
Surrogate Standard Solution 1 (DCAA) (For EPA 515.1)
Catalog No. USPPS1611
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
Catalog No. Quantity
US-PPS-1611 1 x 1 mL
1 options
Catalog No. US-PPS-1611 Supplier Agilent Technologies Supplier No. PPS1611
DCAA Methyl Ester at 100μg/mL in Methyl tert-Butyl Ether
Specifications
Chemical Name or Material Surrogate Standard Solution (DCAA)
Packaging Ampule
Methods For EPA Method 515.1
Quantity 1 x 1 mL