Learn More
Terumo Medical Corporation Surshield-PUR Safety I.V. Catheters 18G
Catalog No. 21200332 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Needle Gauge:
18 G
Catalog No. Needle Gauge
21-200-332 18 G
1 options
This item is not returnable. View return policy
Catalog No. 21-200-332 Supplier Terumo Medical Corporation Supplier No. SRSFF1832A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safety I.V. Catheters 18G x 1 1/4 in.
  • Passive safety to reduce risk of needlestick injury, without adding extra steps to your procedures
  • SurFlash™ enhanced flashback feature provides clear visual confirmation of catheter placement, for increased confidence and first-stick success
  • Easy to use
  • Automatic activation - no extra steps needed
  • A compact design that minimizes the waste volume
Specifications
Material Polyurethane, Polypropylene, Stainless Steel (Needle)
Needle Gauge 18 G
Latex Free Yes
Length (English) 1 1/4 in.
Product Type IV Catheter
Disposable Single-use
For Use With (Equipment) IV Lines
Connection Type Catheter
Graduated No
Sterility Sterile