Learn More
Terumo Medical Corporation Surshield-PUR Safety I.V. Catheters 22G
Catalog No. 21200336 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Length (English):
1 1/4 in.
1 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-200-336 1 1/4 in.
21-200-335 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-200-336 Supplier Terumo Medical Corporation Supplier No. SRSFF2232A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safety I.V. Catheters.
  • Passive safety to reduce risk of needlestick injury, without adding extra steps to your procedures
  • SurFlash™ enhanced flashback feature provides clear visual confirmation of catheter placement, for increased confidence and first-stick success
  • Easy to use
  • Automatic activation - no extra steps needed
  • A compact design that minimizes the waste volume
Specifications
Material Polyurethane, Polypropylene, Stainless Steel (Needle)
Needle Gauge 22 G
Latex Free Yes
Length (English) 1 1/4 in.
Product Type IV Catheter
Disposable Single-use
For Use With (Equipment) IV Lines
Connection Type Catheter
Graduated No
Sterility Sterile