Learn More
Terumo Medical Corporation SurTract™ Retractable Safety Syringe
Catalog No. 22030303 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-030-303 5 mL
22-030-301 1 mL
22-030-302 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-030-303 Supplier Terumo Medical Corporation Supplier No. STR05SS
Add to Cart
Add to Cart
The retractable safety hypodermic line that reduces the risk of needle stick injury through a passive safety system while minimizing patient discomfort during injection.
  • Smooth and comfortable injection due to less plunger pressure
  • Blunt needle allows change of needle for drawing up and prevents needle dullness
  • Reduces medical waste due to lack of external plastic safety sheath
  • Low dead space syringe reduces medication waste
  • Customizable Needle Selection - SurTract™ safety needles are designed for use exclusively with SurTract™ safety syringes allowing you to create the combination you need
Specifications
Material Polypropylene Ethylene Copolymer (Barrel), Polypropylene Ethylene Copolymer (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Retractable Syringe without Needle
Disposable Single-use
Volume (Metric) 5 mL
Connection Type Retractible
Graduated Yes
Sterility Sterile