Learn More
Terumo Medical Corporation SurTract™ Safety Needle 18G (Blunt Fill Needle)
Catalog No. 22030304 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Graduated:
No
Catalog No. Graduated
22-030-304 No
1 options
Catalog No. 22-030-304 Supplier Terumo Medical Corporation Supplier No. STR1832BN
Add to Cart
Add to Cart
The retractable safety hypodermic line that reduces the risk of needle stick injury through a passive safety system while minimizing patient discomfort during injection.
  • Smooth and comfortable injection due to less plunger pressure
  • Blunt needle allows change of needle for drawing up and prevents needle dullness
  • Reduces medical waste due to lack of external plastic safety sheath
  • Low dead space syringe reduces medication waste
  • Customizable Needle Selection - SurTract™ safety needles are designed for use exclusively with SurTract™ safety syringes allowing you to create the combination you need
  • Silicone coating needle outer surface - Polydimethylsiloxane (silicone oil) Medical Grade + Functionalized Siloxane (primer) Medical grade
Specifications
Material Stainless Steel (304), Polypropylene
Needle Gauge 18 G
Latex Free Yes
Product Type Blunt Fill Needle
Disposable Single-use
For Use With (Equipment) SurTract Syringes
Connection Type Fixed
Graduated No
Sterility Sterile