Learn More
Medchemexpress LLC Survodutide TFA | 99.4% | 4231.62 (free base) | 25 MG
SDP
Catalog No. 5000396208
Encompass_Preferred
Survodutide TFA is a potent and selective dual agonist targeting the glucagon receptor (GCGR) and GLP-1 receptor (GLP-1R), with EC50s of 0.52 nM and 0.33 nM respectively in CHO-K1 cells. This 29-amino-acid acylated peptide, featuring a C18 fatty acid, demonstrates significant anti-obesity efficacy by enhancing energy expenditure and reducing food intake.
  • Potent, selective GCGR/GLP-1R dual agonist
  • EC50s of 0.52 nM (GCGR) and 0.33 nM (GLP-1R)
  • 29-amino-acid acylated peptide
  • Contains a C18 fatty acid
  • Demonstrates robust anti-obesity efficacy
  • Increases energy expenditure and decreases food intake
  • Enhanced proteolytic stability
  • Extended terminal half-life
Catalog No. 50-003-96208 Supplier Medchemexpress LLC Supplier No. 25MGHYP4146A
Survodutide TFA is a potent and selective dual agonist targeting the glucagon receptor (GCGR) and GLP-1 receptor (GLP-1R), with EC50s of 0.52 nM and 0.33 nM respectively in CHO-K1 cells. This 29-amino-acid acylated peptide, featuring a C18 fatty acid, demonstrates significant anti-obesity efficacy by enhancing energy expenditure and reducing food intake.
  • Potent, selective GCGR/GLP-1R dual agonist
  • EC50s of 0.52 nM (GCGR) and 0.33 nM (GLP-1R)
  • 29-amino-acid acylated peptide
  • Contains a C18 fatty acid
  • Demonstrates robust anti-obesity efficacy
  • Increases energy expenditure and decreases food intake
  • Enhanced proteolytic stability
  • Extended terminal half-life