Learn More
Sino Biological Anti-IL-10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 62000T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Sus scrofa (Pig) IL-10 (Catalog#62000-WNAE; Q29055; Ser19-Asn175). IL-10 specific IgG was purified by Sus scrofa (Pig) IL-10 affinity chromatography.
Catalog No. 50-241-1869