Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 62003RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Sus scrofa (Pig) IL-1 beta (Catalog#62003-WNAE; NP_999220.1; Ala115-Pro267). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9740