Learn More
Sino Biological Sus scrofa (pig) IL6 / IL-6 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 62006RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Sus scrofa (pig) IL6 / IL-6 (Catalog#62006-WNAE; NP_999564.1; Pro29-Met212). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-8094