Learn More
Sino Biological Sus scrofa (Pig) LGMN Protein 50ug (Sino Biological)
Supplier:  Sino Biological 62001W07H50
Encompass_Preferred
A DNA sequence encoding the sus scrofa(Pig) LGMN (XP_001927117.4) (Val18-Tyr433) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3298