Learn More
Aviva Systems Biology Corp SUSD3 Antibody - N-terminal region (ARP55443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUSD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SUSD3. Alternate names: MGC26847ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-53755