Learn More
Aviva Systems Biology Corp SUSD4 Antibody - middle region (ARP47384_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47384_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUSD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUSD4. Alternate names: FLJ10052, PRO222, RP11-239E10.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59533