Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Suspended Pencil Drawer Painted Steel Cabinet
GREENER_CHOICE

Catalog No. 06000514 Shop All Fisherbrand Products
Change view
Click to view available options
Suspended Pencil Drawer Painted Steel Cabinet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
06-000-514 Suspended Pencil Drawer Painted Steel Cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-000-514
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fisherbrand™ Suspended Pencil Drawer Painted Steel Cabinets are durable and constructed with SEFA 8 Metal compliant load-bearing capacity.

  • Suspended Cabinets are designed to be hung from any of the Fisherbrand Table Systems.
  • Each suspended cabinet is furnished with complete hanging hardware.
  • Fisherbrand White is a urethane powder coating that provides reliable chemical and abrasion resistance.

Specifications

Color Fisherbrand White
Includes Hanging Hardware
For Use With (Equipment) Fisherbrand Table Systems
Depth (English) 22 in.
Depth (Metric) 55.9 cm
Height (English) 3.86 in.
Height (Metric) 9.8 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.