Learn More
IPHASE PHARMA SERVICES LLC Suspension Beagle Dog Hepatocytes,Male,5million
Catalog No. NC3966918
Encompass_Preferred
Primary Hepatocytes are isolated from human and animal liver tissues. It's a tool for in vitro metabolic stability and hepatocyte metabolic stability. IPHASE provides both suspension and plateable Hepatocytes, animal or human and speical species includes: cat(feline), Guinea pig, BALB/c Nude, Ferret, Gerbil customize hepatocytes. Suspension Hepatocytes need to be used in conjunction with Thaw medium and Incubation medium. These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3966918 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 01931C1.21
Primary Hepatocytes are isolated from human and animal liver tissues. It's a tool for in vitro metabolic stability and hepatocyte metabolic stability. IPHASE provides both suspension and plateable Hepatocytes, animal or human and speical species includes: cat(feline), Guinea pig, BALB/c Nude, Ferret, Gerbil customize hepatocytes. Suspension Hepatocytes need to be used in conjunction with Thaw medium and Incubation medium. These products require shipment in LN2 container therefore customer need to return the container.