Learn More
Med Vet International Ethicon Suture Chromic Gut 3-0 SH 36/Box G122H
Catalog No. NC1985420
Encompass_Preferred
The Ethicon G122H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a SH taper point needle and has 36 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1985420 Supplier Med Vet International Supplier No. G122H
The Ethicon G122H suture is a high-quality absorbable Chromic Gut surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed brown to provide greater visibility while suturing, and typically have an absorption rate of 90 days. This suture comes with a SH taper point needle and has 36 sutures in a box.